Orders LinkContactUsLinkComponent
Cyclic Olefin, Reference Standards
Cyclic Olefin, Reference Standards
Leverancier:   USP
Cyclic Olefin, Reference Standards
Leverancier:  USP
Certificaten
2 Opties Hieronder Beschikbaar
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certificeringen: US Pharmacopea Reference Standard
Specifications for Product Family
Specificaties
Specificaties
  • Catalog No:
  • 1152814.
  • 1152825.
  • Omschrijving:
  • Cyclic Olefin Polymer
  • Cyclic Olefin Copolymer
  • CAS No.:
  • 26007-43-2
Specificaties
Product Family Options
Productinformatie
  • Standaard
    Info VPE
    Verpakking
    VPE
    Beschikbaarheid
    Price
  • 1152814.
    Cyclic Olefin Polymer
    Cyclic Olefin Polymer
    USP reference standard
    Vial
    1 g
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 1 g
    1 * 1 g - 416,00 €
    416,00 €
    1 * 1 g
       416,00 €
    Specificaties:
    More
    Specifications
    Cat. No.
    1152814.
    Omschrijving
    Cyclic Olefin Polymer
    CAS No.
  • 1152825.
    Cyclic Olefin Copolymer
    Cyclic Olefin Copolymer
    USP reference standard
    3 strips, 2'' x 2'' x 1 mm each
    1 g
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 1 g
    1 * 1 g - 424,00 €
    424,00 €
    1 * 1 g
       424,00 €
    Specificaties:
    More
    Specifications
    Cat. No.
    1152825.
    Omschrijving
    Cyclic Olefin Copolymer
    CAS No.
    26007-43-2
Aanbevelingen
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.