4-Methoxybenzenediazonium tetrafluoroborate 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:459-64-3
:
Formula:C₇H₇BF₄N₂O
MDL Number:MFCD00011897
Molecular Weight:221.95 g/mol
Melting Pt:142…144 °C
Storage Temperature:Kamertemperatuur
EINECS:207-296-2
UN:2923
ADR:8 III
:GHS05
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay Minimum:98
Specification Test Results
Related Information
Frequently Bought Together