Burkle 0006-4002 Threaded Adapter for Mini Solvent Pump, ETFE; GL45-GL38
Catalog # MFLX06040-02
Supplier: Avantor Fluid Handling
Specifications
- Omschrijving:Threaded Adapter for Mini Solvent Hand Pump, ETFE; GL45-GL38
- Wetted materials:ETFE
- Garantie:15 Months
- VPE:1
- Container adapter:GL38
Specifications
About this item
Suitable for use with Hand-Operated Mini Solvent Pumps.
- Order optional adapters to fit a wide range of containers
- Avaiable in different material and thread configurations
- Withstand temperatures up to 104 °F (40 °C)
The 06043-95 hand pump features a GL45 connecting thread made of PTFE for connection to a wide range of containers. Optional S40, GL38 and GL32 adapters in PTFE or ETFE are available separately to easily adapt this hand operating pump to other bottles, canisters, or tanks.
- Omschrijving:Threaded Adapter for Mini Solvent Hand Pump, ETFE; GL45-GL38
- Environmentally Preferable:
- Wetted materials:ETFE
- Garantie:15 Months
- VPE:1
- Container adapter:GL38