Compression Elbow Threaded Adapter, Stainless Steel, 1/4" x 1/4", Each
Catalog # MFLX31406-78
Supplier: Avantor Fluid Handling
CE certificate
Specifications
- Verbindingstype:Compression to Thread
- Flow pattern:Elbow
- Thread size NPT(M) (port 2):1/4"
- Omschrijving:Compression Elbow Threaded Adapter, Stainless Steel, 1/4"×1/4", Each
- Sterilisatie:Autoclave, ethylene oxide (ETO), or gamma
- Garantie:LTD life time
- VPE:1
- Fitting material:Stainless Steel 316
Specifications
About this item
Create a secure connection with compression fittings.
- Constructed of durable stainless steel (SS)
Very good chemical resistance. Pressure limitation determined by the tubing, not the fitting. Usable from -40 to 200ºF (-40 to 93ºC).
- Verbindingstype:Compression to Thread
- Flow pattern:Elbow
- Thread size NPT(M) (port 2):1/4"
- Omschrijving:Compression Elbow Threaded Adapter, Stainless Steel, 1/4"×1/4", Each
- Environmentally Preferable:
- Sterilisatie:Autoclave, ethylene oxide (ETO), or gamma
- Garantie:LTD life time
- VPE:1
- Fitting material:Stainless Steel 316