Corning® 25-Layer CellCube® Module, Corning
Over dit artikel
The CellCube Module is an integral, encapsulated, sterile, single-use device that is 100% pressure-hold tested before shipment.
It is comprised of a series of parallel, styrene plates joined to create thin, sealed, laminar flow spaces between adjacent plates. Each culture plate receives proprietary tissue culture treatment prior to assembly to ensure dense, uniform cell growth. These CellCube Modules provide: a series of parallel, polystyrene plates joined to create thin, sealed, laminar flow spaces between adjacent plates, 21,250 cm² polystyrene growth surface optimally treated for the growth of attachment dependent cells, a perfused environment which more closely simulates in vivo conditions and reliably distributes nutrients and oxygen, an integral, encapsulated, sterile, single-use device that is 100% pressure-hold tested before shipment. Square Flask Shape.
3 Opties Hieronder Beschikbaar
Product Details & Documents
The CellCube Module is an integral, encapsulated, sterile, single-use device that is 100% pressure-hold tested before shipment.
It is comprised of a series of parallel, styrene plates joined to create thin, sealed, laminar flow spaces between adjacent plates. Each culture plate receives proprietary tissue culture treatment prior to assembly to ensure dense, uniform cell growth. These CellCube Modules provide: a series of parallel, polystyrene plates joined to create thin, sealed, laminar flow spaces between adjacent plates, 21,250 cm² polystyrene growth surface optimally treated for the growth of attachment dependent cells, a perfused environment which more closely simulates in vivo conditions and reliably distributes nutrients and oxygen, an integral, encapsulated, sterile, single-use device that is 100% pressure-hold tested before shipment. Square Flask Shape.
Documents
Certificaten
Zoeken op lotnummerSpecifications for Product Family
Specificaties
- Catalog No:
- CORN3201
- CORN3304
- CORN3241
- Environmentally Preferable:
- Lage impact op de fabricage
- Mediavolume:
- 1500 ml
- 1500 ml
- Oppervlakte:
- Tissue Culture Treated
- Corning® CellBIND®
- Milieu certificering:
- RECs, ISO 14001 - Kennebunk
Specificaties
Product Family Options
Productinformatie
- CultuuroppervlakteBeschikbaarheidPrice
- CORN320125-layer CellCube® module with 21,250 cm² growth surface, Polycarbonate/Polystyrene, Sterile25-layer CellCube® module with 21,250 cm² growth surface, Polycarbonate/Polystyrene, Sterile21,250 cm²Log in voor beschikbaarheid1 * 1 ST698,00 €Specificaties:
Lage impact op de fabricage More
Specifications
Cat. No.CORN3201Environmentally PreferableLage impact op de fabricageMediavolume1500 mlOppervlakteTissue Culture TreatedMilieu certificeringRECs, ISO 14001 - KennebunkLage impact op de fabricage - CORN330425-layer CellCube® module with 21,250 cm² growth surface, Polycarbonate/Polystyrene, Sterile25-layer CellCube® module with 21,250 cm² growth surface, Polycarbonate/Polystyrene, Sterile21,250 cm²Log in voor beschikbaarheid1 * 1 ST1 430,00 €Specificaties:
More
Specifications
Cat. No.CORN3304Environmentally PreferableMediavolume1500 mlOppervlakteCorning® CellBIND®Milieu certificering - CORN3241Corning CellCube®, 25-layer, CellBIND® Surface, Polystyrene with 6"Corning CellCube®, 25-layer, CellBIND® Surface, Polystyrene with 6"Log in voor beschikbaarheid1 * 5 ST913,00 €Specificaties:
More
Specifications
Cat. No.CORN3241Environmentally PreferableMediavolumeOppervlakteMilieu certificering