ITT Jabsco Polypropylene Pulse Dampeners With Santoprene Diaphragm, 3/8" NPT
Catalogus # MFLX74021-04
Leverancier: Avantor Fluid Handling
Specificaties
- Omschrijving:Polypropylene Pulse Dampeners With Santoprene Diaphragm, 3/8" NPT
- VPE:1
- Wetted materials:Polypropylene, Santoprene®
- Luchtinlaataansluiting:0,25 NPT(F)
- Inlet connection:0,375 NPT(F)
- Outlet connection:0,375 NPT(F)
- Garantie:1 year
Specificaties
Over dit artikel
Create a pulse-free flow out of your positive displacement pump.
- Choose between PTFE or Polypropylene with Santoprene dampners
Dampeners work by pre-filling the chamber with compressed air or other gas, then as fluid pressure increases it compresses the air/gas in the chamber. When the air/gas pressure exceeds the fluid pressure the fluid is forced back out of the dampener in between pulse cycles, resulting in a near pulseless flow.
More Info: These pulse dampeners can be used with any positive displacement pump that needs flow with reduced pulsation.
- Omschrijving:Polypropylene Pulse Dampeners With Santoprene Diaphragm, 3/8" NPT
- Environmentally Preferable:
- VPE:1
- Wetted materials:Polypropylene, Santoprene®
- Luchtinlaataansluiting:0,25 NPT(F)
- Inlet connection:0,375 NPT(F)
- Outlet connection:0,375 NPT(F)
- Garantie:1 year