Pressure Relief Valve, 316 SS, 1/2" NPT(F)
MFLX03245-40
- Verbindingstype:NPT(F)
- Max. druk:150 psi
- Omschrijving:Pressure Relief Valve, 316 SS, 1/2" NPT(F)
- Wetted materials:Stainless Steel
- Garantie:1 year
- Orifice:7,9375 mm (0,3125")
- VPE:1
Easily control your outlet pressure.
- Constructed of durable 316 stainless steel (SS)
Use for liquid applications with flow rates up to 15 GPM. For pressures from 40 to 150 psig. Valves have 1/2" NPT(F); orifice size is 5/16".
- Verbindingstype:NPT(F)
- Max. druk:150 psi
- Omschrijving:Pressure Relief Valve, 316 SS, 1/2" NPT(F)
- Environmentally Preferable:
- Wetted materials:Stainless Steel
- Garantie:1 year
- Orifice:7,9375 mm (0,3125")
- VPE:1