Bel-Art Flow Control Clamp, Delrin Screw Clamp, 3/Pk
MFLX06833-10
- Omschrijving:Bel-Art Flow Control Clamp, Delrin Screw Clamp, 3/Pk
- Klemmateriaal:Delrin acetal
- VPE:3
- Klemtype:Tubing
- Max. grip range:0,5"
Deliver accurate regulation or interruption of fluid flow.
- Constructed of Delrin acetal
DELRIN(R) resists weak acids, bases, and organic solvents. Controls flow in flexible tubing up to 1/2" (13 mm) OD. Replaces stopcocks in many applications. Install without disconnecting tubing. Autoclavable. Bel-Art.
- Omschrijving:Bel-Art Flow Control Clamp, Delrin Screw Clamp, 3/Pk
- Environmentally Preferable:
- Klemmateriaal:Delrin acetal
- VPE:3
- Klemtype:Tubing
- Max. grip range:0,5"