CPC® Miniature Quick-Disconnect Fittings, Panel-Mount Hose Barb Inserts
Supplier: Avantor Fluid Handling
Certificates
About this item
No more kinked tubing—allows tubing to rotate freely.
- Twist-to-connect couplings provide a reliable and more secure alternative to luer-type connections
- Tubing can rotate freely when connected—preventing kinked tubing and accidental disconnection
These CPC® miniature quick-disconnect couplings are able to handle 1/16" ID tubing, and offer a more secure connection than luer fittings. Fitting halves engage and seal with a twist. Once engaged, the coupling allows the tubing to rotate freely.
For a complete coupling, order one fitting body and one insert.
- Environmentally Preferable:
Specifications
- Maximumtemperatuur:
- 82 °C (180 °F)
- 82 °C (180 °F)
- 82 °C (180 °F)
- Min. temperatuur:
- –40 °C (–40 °F)
- –40 °C (–40 °F)
- 0 °C (32 °F)
- O-Ring material:
- Buna N
- Buna N
- EPDM
- Sterilisatie:
- Autoclave or ethylene oxide (ETO)
- Autoclave or ethylene oxide (ETO)
- Ethylene oxide (ETO) or gamma
- Valve material:
- None
- None
- None
- Garantie:
- 1 year
- 3 Months
- 3 Months
Specifications
Frequently Bought Together