Orders LinkContactUsLinkComponent
Dumont #4 forceps
Dumont #4 forceps
Marketsource
Supplier:   FINE SCIENCE TOOLS
Dumont #4 forceps
Supplier:  FINE SCIENCE TOOLS
Marketsource
Certificates
Dumont #4 forceps
Dumont #4 forceps
2 Options Available Below
Product Details & Documents
Product Family Options
Product Information
  • Type
    Materiaal
    Lengte
    Tip stijl
    Tip shape
    Tipgrootte
    Availability
    Price
  • #4
    Dumoxel
    110 mm
    Biology
    Straight
    0,06×0,04 mm
    Log in for availability
    Log in for availability
    1 * 1 Pce
    1 * 1 Pce - 38,50 €
    38,50 €
    1 * 1 Pce
       38,50 €
    Specifications:
    Marketsource
    Marketsource
  • #4
    Dumoxel
    110 mm
    Standard
    Straight
    0,13×0,08 mm
    Log in for availability
    Log in for availability
    1 * 1 Pce
    1 * 1 Pce - 35,50 €
    35,50 €
    1 * 1 Pce
       35,50 €
    Specifications:
    Marketsource
    Marketsource