Dumont #5CO forceps
#5CO, Dumostar, 110 mm, Biology, Straight, 0,05×0,01 mm
Catalogus # FSCI11295-20
Marketsource
Leverancier: FINE SCIENCE TOOLS
Ga naar:
Product Details & Documents
Specificaties
- Type:#5CO
- Lengte:110 mm
- Materiaal:Dumostar
- Tip stijl:Biology
- Tipgrootte:0,05×0,01 mm
- Tip shape:Straight