Dumont #7b forceps
#7b, Inox, 110 mm, Standard, Curved, 0,17×0,1 mm
Catalogus # FSCI11270-20
Marketsource
Leverancier: FINE SCIENCE TOOLS
Ga naar:
Product Details & Documents
Specificaties
- Type:#7b
- Lengte:110 mm
- Materiaal:Inox
- Tip stijl:Standard
- Tipgrootte:0,17×0,1 mm
- Tip shape:Curved