Dumont mini forceps, style 3
Mini #3, Inox, 70 mm, Standard, Straight, 0,12×0,07 mm
Catalogus # FSCI11200-10
Marketsource
Leverancier: FINE SCIENCE TOOLS
Ga naar:
Product Details & Documents
Specificaties
- Type:Mini #3
- Lengte:70 mm
- Materiaal:Inox
- Tip stijl:Standard
- Tipgrootte:0,12×0,07 mm
- Tip shape:Straight