Orders LinkContactUsLinkComponent
Round handled needle holders
Round handled needle holders
Marketsource
Supplier:   FINE SCIENCE TOOLS
Round handled needle holders
Supplier:  FINE SCIENCE TOOLS
Marketsource
Certificates
Round handled needle holders
Round handled needle holders
2 Options Available Below
Product Details & Documents
Product Family Options
Product Information
  • Omschrijving
    Type
    Materiaal
    Lengte
    Tip shape
    Tip width
    Availability
    Price
  • Clamping length of 7 mm
    Lockable
    Roestvrij staal
    125 mm
    Straight
    0,6 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 278,00 €
    278,00 €
    1 * 1 ST
       278,00 €
    Specifications:
    Marketsource
    Marketsource
  • Clamping length of 7 mm
    Roestvrij staal
    125 mm
    Straight
    0,6 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 257,00 €
    257,00 €
    1 * 1 ST
       257,00 €
    Specifications:
    Marketsource
    Marketsource