Orders LinkContactUsLinkComponent
Double micro clamps
Double micro clamps
Marketsource
Supplier:   FINE SCIENCE TOOLS
Double micro clamps
Supplier:  FINE SCIENCE TOOLS
Marketsource
Certificates
Double micro clamps
Double micro clamps
3 Options Available Below
Product Details & Documents
Product Family Options
Product Information
  • Omschrijving
    Materiaal
    Lengte
    Tip shape
    Tipgrootte
    Availability
    Price
  • Variable clamp press, 5 - 15 g
    Roestvrij staal
    7 mm
    Straight
    3,5×1 mm
    Log in for availability
    Log in for availability
    1 * 1 Pce
    1 * 1 Pce - 257,50 €
    257,50 €
    1 * 1 Pce
       257,50 €
    Specifications:
    Marketsource
    Marketsource
  • Variable clamp press, 5 - 15 g
    Roestvrij staal
    11 mm
    Straight
    5,5×1,5 mm
    Log in for availability
    Log in for availability
    1 * 1 Pce
    1 * 1 Pce - 253,50 €
    253,50 €
    1 * 1 Pce
       253,50 €
    Specifications:
    Marketsource
    Marketsource
  • Variable clamp press, 5 - 15 g
    Roestvrij staal
    16 mm
    Straight
    7,5×1,75 mm
    Log in for availability
    Log in for availability
    1 * 1 Pce
    1 * 1 Pce - 244,50 €
    244,50 €
    1 * 1 Pce
       244,50 €
    Specifications:
    Marketsource
    Marketsource