Orders LinkContactUsLinkComponent
Imager Tissue Package, THUNDER
Imager Tissue Package, THUNDER
Leica® THUNDER imager tissue package
Catalogus # 630-3395
Leverancier:   LEICA MICROSYSTEMS
Imager Tissue Package, THUNDER
Catalogus # 630-3395
Leverancier:  LEICA MICROSYSTEMS
Imager Tissue Package, THUNDER
Imager Tissue Package, THUNDER
Leica® THUNDER imager tissue package
Price
99 782,75 €
Log in voor beschikbaarheid
1 * 1 SET
Product Details & Documents
The THUNDER imager tissue allows real time fluorescence imaging of 3-D tissue sections typically used in neuroscience and histology research. Acquire rich, detailed images of thick tissues free of haze from ‘out of focus’ blur.
Certificeringen: CE and CAS.
Levering: THUNDER Imager Tissue Imager Package includes: DM4 B semi-motorised upright microscope stand, manual camera port, motorised 5-position fluorescence filter turret, coded 7-position objective nosepiece, objectives (Plan FL 1,25×/0,04 N.A., Plan FL 10×/0,32 N.A. and Plan Apo 40×/0,95 N.A. with correction collar), 10×/25 mm FOV eyepieces, LED transmitted light, brightfield condenser (0,90 N.A.), manual stage (110° rotation), shutter controlled LED3 fluorescence light source, fluorescence filters (LED405, GFP and RHOD), K5 sCMOS camera; dust cover, THUNDER Workstation 2D, US keyboard, LAS X Core software, LAS X Multi-Channel Acquisition software module, 32" monitor and 1 TB SSD RAID addition.
Purchase of THUNDER Imager Tissue Package includes service installation and 1 day of applications training by an LMS Advanced Workflow Specialist.
Specificaties
  • Omschrijving:
    Leica® THUNDER imager tissue package
  • Seq:
    01
Aanbevelingen
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.