ContactUsLinkComponent
Comfort Plus™ Industrial safety helmet, EVO®3
Comfort Plus™ Industrial safety helmet, EVO®3
Catalog # JSPAAJF160-000-100
Supplier:  JSP
undefined
Comfort Plus™ Industrial safety helmet, EVO®3
Catalog # JSPAAJF160-000-100
Supplier:  JSP
Specifications
  • Omschrijving:
    EVO®3 Industrial safety helmet
  • VPE:
    1
Specifications
About this item
The EVO®3 industrial safety helmet combines a super strong shell for superior all day protection, with the comfort benefits of the Evolution® 3D Adjustment™ harness system
  • Industrial safety helmet
  • Tough HDPE shell
  • 6-point terylene cradle harness
  • Prescision fit
  • Vented design
  • OneTouch™ slip ratchet adjustment
The EVO®3 Comfort Plus™ helmet combines a super strong shell for superior all day protection in the widest range of environments, with the comfort benefits of the Evolution® 3D Adjustment™ harness system. In tests the EVO®3 proved far tougher than the EN 397 standard required and far tougher than any of its competitors.
A 6-point terylene cradle harness system offers unrivalled comfort without compromising performance. Egyptian cotton core with porous PU coating for maximum sweat absorption. pH neutral, dermatologically tested. Never before has such a precise fit been attainable on an industrial safety helmet, using the unique 1-2-3 point harness depth settings.
Certifications: EN 397.
Eyewear for safety helmets
Product Information