ContactUsLinkComponent
Comfort Plus™ industrial safety helmet, EVO®2
Comfort Plus™ industrial safety helmet, EVO®2
Catalogus # JSPAAJF030-000-100
Leverancier:  JSP
undefined
Comfort Plus™ industrial safety helmet, EVO®2
Catalogus # JSPAAJF030-000-100
Leverancier:  JSP
Specificaties
  • Omschrijving:
    EVO®2 Industrial safety helmet
  • VPE:
    1
Specificaties
Over dit artikel
The EVO®2 offers the most economical top level protection, combining a tough HDPE shell with a 6-point polyethylene cradle harness for unrivalled comfort.
  • Industrial safety helmet
  • Tough HDPE shell
  • 6-point polypropylene cradle harness
  • Prescision fit
  • Vented design
  • OneTouch™ slip ratchet adjustment
The EVO®2 Comfort Plus™ helmet combines a super strong shell for superior all day protection in the widest range of environments, with the comfort benefits of the Evolution® 3D Adjustment™ harness system. In tests the EVO®2 proved far tougher than the EN 397 standard required and far tougher than any of its competitors.
A 6-point polyethylene cradle harness system offers unrivalled comfort without compromising performance. Egyptian cotton core Chamlon sweatband with porous PU coating for maximum sweat absorption. pH neutral, dermatologically tested. Never before has such a precise fit been attainable on an industrial safety helmet, using the unique 1-2-3 point harness depth settings.
Certificeringen: EN 397.