Double lens goggle, EVO®
EVO® double lens goggle, PC outer lens, Acetate inner lens
Catalogus # JSPAAGM020-823-000
Leverancier: JSP
Ga naar:
Product Details & Documents
Double lens goggle engineered to offer outstanding performance, compatibility, comfort, and looks.
- Double lens design
- Indirect venting
- PVC frame
- Elastic headband
- Contour design
- High level of protection
- Fully compatible with the JSP EVO® Range of industrial safety helmets
Double lens design for optimum anti fog protection, as well as anti-mist and scratch resistant lens with UV protection. Low profile design is lightweight with soft EVA foam housing for exceptional wearer comfort. Googles provide dust, liquid splash, molten metal, and extreme temperature protection. Compatible with prescription lenses by using RX Inserts for EVO® Goggles.
Certificeringen: EN 166; EN 170.
Documents
Manual Safety Eyewear
Handmatig
TDS Goggles Evo Double Lens
Technische fiche
Specificaties
- Omschrijving:EVO® double lens goggle
- Beglazing:PC outer lens, Acetate inner lens
Accessories
Productinformatie
- Catalogus #BeschikbaarheidPrice
- RX prescription insert for EVO® goggles, TR 90 material, 6 g, 6 mm curveRX prescription insert for EVO® goggles, TR 90 material, 6 g, 6 mm curveJSPAAGU230-001-300Log in voor beschikbaarheid1 * 1 ST - 5,80 €