ContactUsLinkComponent
Wall mounting bracket for waste pipes, b.safe
Wall mounting bracket for waste pipes, b.safe
Catalog # BOHLR1164-03
Supplier:  Bohlender
undefined
Wall mounting bracket for waste pipes, b.safe
Catalog # BOHLR1164-03
Supplier:  Bohlender
Specifications
  • Omschrijving:
    Wall mounting bracket for waste pipes, L×W: 260×44 mm
  • VPE:
    1
Specifications
About this item
Black, powder-coated plate made of steel. With 2 bores Ø 4,2 mm for wall mounting with the included fixing material.
  • Compact collective connector to combine the outlets of two respectively up to four HPLC units
  • All connectors are placed sideways to join tubing and tubes with suitable fittings and hose connectors without bending
Waste Pipes with tube Ext.Ø 32 mm can be fixed and adjusted in height on the long side of the plate by using suitable b.safe Magnet Holders.