Elbow stopcock, b.safe
Catalog # BOHLR1201-05
Supplier: Bohlender
Specifications
- Boring Ø:10 mm
- Schroefdraad:GL 25 (Male and female)
- VPE:1
Specifications
About this item
90° Elbow fitting. To connect b.safe Waste Tubes to b.safe Waste Caps without bending. Integrated stopcock with plug made of PTFE-EX to stop the flow through b.safe Waste Tubes when canisters are to be exchanged or emptied.
- Made of static dissipative PE-EX
- Firmly mounted union nut made of PPS-EX with female thread GL 25
- Outer dimension: 34×52×102 mm
To connect b.safe Waste Tubes to b.safe Waste Caps GL 25 without bending. No dripping from b.safe Waste Tubes during exchange of collection canisters.