Orders LinkContactUsLinkComponent
Nitrogen generator, Precision Nitrogen Headspace 250
Nitrogen generator, Precision Nitrogen Headspace 250
Precision Nitrogen Headspace 250 nitrogen generator
Catalogus # 564-0298
Leverancier:   PEAK SCIENTIFIC
Nitrogen generator, Precision Nitrogen Headspace 250
Catalogus # 564-0298
Leverancier:  PEAK SCIENTIFIC
Nitrogen generator, Precision Nitrogen Headspace 250
Nitrogen generator, Precision Nitrogen Headspace 250
Precision Nitrogen Headspace 250 nitrogen generator
Price
5 990,00 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
The Precision Nitrogen Headspace 250 has been conceived and custom-engineered to meet the gas requirements for headspace sampler instruments requiring higher gas pressure.
  • Ultra High Purity nitrogen for headspace sampler instruments
  • Can supply make-up gas
  • Single unit can stack with the Precision Hydrogen, Nitrogen and Zero Air generators
  • Service and repair status indicators
  • 1 year comprehensive on-site warranty
Specificaties
  • Omschrijving:
    Precision Nitrogen Headspace 250 nitrogen generator
  • Cat. No.:
    564-0298