Cellulose chromatography and blotting paper roll, Whatman™
N° de catalogue WHAT10539028
Fournisseur: Whatman products (Cytiva)
Spécifications
- Maat:3.8 cm × 137.1 m
- VPE:1
Spécifications
À propos de cet article
Cellulose Chromatography and Blotting Paper Roll contains cotton linter filter sheets for electrophoresis and amino acid chromatography.
It is also ideal for use with gelatinous samples.