Orders LinkContactUsLinkComponent
Spring scissors, ceramic coated
Spring scissors, ceramic coated
Marketsource
Supplier:   FINE SCIENCE TOOLS
Spring scissors, ceramic coated
Supplier:  FINE SCIENCE TOOLS
Marketsource
Certificates
Spring scissors, ceramic coated
Spring scissors, ceramic coated
2 Options Available Below
Product Details & Documents
These scissors are made of ceramic coated stainless steel.
Product Family Options
Product Information
  • Type
    Lengte
    Lengte blad
    Tipgrootte
    Availability
    Price
  • Sharp, curved tips
    100 mm
    7 mm
    0,1 mm
    Log in for availability
    Log in for availability
    1 * 1 Pce
    1 * 1 Pce - 368,50 €
    368,50 €
    1 * 1 Pce
       368,50 €
    Specifications:
    Marketsource
    Marketsource
  • Sharp, straight tips
    100 mm
    7 mm
    0,1 mm
    Log in for availability
    Log in for availability
    1 * 1 Pce
    1 * 1 Pce - 339,00 €
    339,00 €
    1 * 1 Pce
       339,00 €
    Specifications:
    Marketsource
    Marketsource