Flat bottom tubes, glass, Assistent®
50×15 mm Ø, 1,2 mm
Catalogus # HECH92277912400
Leverancier: GLASWARENFABRIK KARL HECHT
Flat bottom tubes, glass, Assistent®
Catalogus # HECH92277912400
Leverancier: GLASWARENFABRIK KARL HECHT
Ga naar:
Product Details & Documents
AR-Glas®, special size.
Specificaties
- Wanddikte:1,2 mm
- Maat:50×15 mm Ø