Orders LinkContactUsLinkComponent
QUANTOFIX Copper Sensitive 5
QUANTOFIX Copper Sensitive 5
QUANTOFIX Copper Sensitive 5, 0 • 0.1 • 0.5 • 1.0 • 2.0 • 5.0 - mg/L - Cu⁺ ²⁺, Complexation with biquinoline, 40×40×112 mm (package), 25 test(s), 25
Catalogus # MANA91358
Leverancier:   MACHEREY-NAGEL
QUANTOFIX Copper Sensitive 5
Catalogus # MANA91358
Leverancier:  MACHEREY-NAGEL
CAS-nummer:  
QUANTOFIX Copper Sensitive 5
QUANTOFIX Copper Sensitive 5
QUANTOFIX Copper Sensitive 5, 0 • 0.1 • 0.5 • 1.0 • 2.0 • 5.0 - mg/L - Cu⁺ ²⁺, Complexation with biquinoline, 40×40×112 mm (package), 25 test(s), 25
CAS-nummer:  
Price
14,90 €
Log in voor beschikbaarheid
1 * 25 ST
Product Details & Documents
Semi-quantitative test strips for reliable determination of copper concentrations as low as 0.1 mg/L (ppm). A hole in the plastic carrier allows the water to migrate through the test pad increasing the sensitivity significantly. Ideal for rapid and easy analysis of copper directly on-site. No maintenance or calibration is required.
  • Sensitive rapid test
Specificaties
Specificaties
  • Chemische methode:
    Complexation with biquinoline
  • VPE:
    25
  • Omschrijving:
    QUANTOFIX Copper Sensitive 5
  • Afmetingen:
    40×40×112 mm (package)
  • Tests per kit:
    25 test(s)
  • Meetbereik:
    0 • 0.1 • 0.5 • 1.0 • 2.0 • 5.0 - mg/L - Cu⁺ ²⁺
  • Cat. No.:
    MANA91358
  • Platform:
    Semi-quantitative test strips
  • Instrument:
    No
  • Parameter:
    All reagents included
  • Method:
    Copper - organic complexation (white → red-violet)
  • Bestanddelen:
    25 test strips in a tube, instruction leaflet
  • Gewicht:
    31 g
  • Houdbaarheid (in maanden):
    30 (from production)
Specificaties
Aanbevelingen
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.