Orders LinkContactUsLinkComponent
Solid phase extraction cartridges, Bond Elut™ CN-U
Leverancier:   VARIAN
Solid phase extraction cartridges, Bond Elut™ CN-U
Leverancier:  VARIAN
Certificaten
Solid phase extraction cartridges, Bond Elut™ CN-U
4 Opties Hieronder Beschikbaar
Product Details & Documents
Bond Elut CN-U is ideally suited for the extraction of polar compounds from a nonpolar matrix, such as hexane or oils. It is suitable for very polar analytes that may be irreversibly retained on SI or Diol (2OH) SPE phases.
  • Ideal for extracting aqueous analytes
  • Retention in aqueous and organic matrices
  • Useful for many applications
A medium polarity sorbent with many uses, it is ideal for applications in which extremely nonpolar compounds would be irreversibly retained on high carbon load sorbents, such as C8 and C18. This endcapped version of the cyano sorbent is best utilized when extracting analytes from an aqueous matrix.
Product Family Options
Bond Elut™ Jr cartridge
Productinformatie
  • Fase
    Partikelgrootte
    Capaciteit
    Bedgewicht
    Beschikbaarheid
    Price
  • VARI12166053B
    Bond Elut™ Jr cartridge
    Bond Elut™ Jr cartridge
    Cyano
    40 µm
    1000 mg
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 100 ST
    1 * 100 ST - 432,00 €
    432,00 €
    1 * 100 ST
       432,00 €
LRC cartridges
Productinformatie
  • Fase
    Partikelgrootte
    Capaciteit
    Bedgewicht
    Beschikbaarheid
    Price
  • VARI12113034
    LRC cartridge
    LRC cartridge
    Cyano
    40 µm
    10 ml
    500 mg
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 50 ST
    1 * 50 ST - 243,00 €
    243,00 €
    1 * 50 ST
       243,00 €
Straight barrel cartridges
Productinformatie
  • Fase
    Partikelgrootte
    Capaciteit
    Bedgewicht
    Beschikbaarheid
    Price
  • VARI12102066
    Straight barrel cartridge
    Straight barrel cartridge
    Cyano
    40 µm
    1 ml
    50 mg
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 100 ST
    1 * 100 ST - 276,00 €
    276,00 €
    1 * 100 ST
       276,00 €
  • 551-0304
    Straight barrel cartridge
    Straight barrel cartridge
    Cyano
    40 µm
    1 ml
    100 mg
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 100 ST
    1 * 100 ST - 272,00 €
    272,00 €
    1 * 100 ST
       272,00 €