HRP-conjugate stabiliser, SeramunStab® STNP
Catalog # APOSBIL3113-100ML
Supplier: Apollo Scientific
CAS Number:
Specifications
- VPE:100 mL
- Omschrijving:SeramunStab® STNP HRP-conjugate stabiliser
- Seq:01
Specifications
About this item
Liquid product red odourless pH 6,7 to 6,9. Protein free Stabiliser. Enzyme activity preservation by special chemistry.