- VPE:1
- Omschrijving:SideStep lysis and stabilization buffer, Cell lysis and immediate stabilization of RNA and DNA for real-time PCR analysis. Contains 10 ml buffer
- Environmentally Preferable:
- Info VPE:100 reactions
Progress from mammalian cells to qPCR or QRT-PCR experiments without nucleic acid purification.
- The optimised cell lysis solution stabilises RNA for at least 6 months
- Samples are prepared ready for QRT-PCR analysis in minutes
- The product ensures minimal RNA loss during preparation and analysis
- The buffer employs a single-tube, solution-based format, rather than a column-based format
- The process involves a single PBS wash followed by cell lysis in the SideStep lysis buffer
- Immediately stabilise nucleic acid without further processing required, for more efficient analyses
The Agilent SideStep II Lysis and Stabilisation Buffer uses a single-tube, solution-based format, rather than a column-based format. The patent-pending solution lyses cells in minutes and releases both RNA and DNA molecules into the solution. This immediately stabilises nucleic acids without further processing required. The Cell Lysis and Protein Extraction Kit prevents sample loss and degradation, ensuring accurate gene quantification in downstream QRT-PCR. Large numbers of mammalian samples can be analysed faster and more easily using the buffer, with minimal RNA loss. Skip nucleic acid purification steps in your applications today.
Our SideStep Lysis and Stabilisation Buffer ensures accurate gene expression analysis by eliminating sample loss and degradation. The proprietary buffer lyses cells and immediately stabilises the released nucleic acid for months. Several formats are available to enhance research versatility. Following a 10 minute room temperature lysis step, you can go directly from cells to qPCR amplification without stopping to purify. Advanced screening saves time and money by selecting the best RNA samples for purification. In addition, with a slight protocol modification the buffer can easily be used for detecting miRNA in cell lysates.