Single tool holster with retractor, DBI-SALA, 3M™
Catalog # CPSD1500104
Supplier: CAPITAL SAFETY
Specifications
- Omschrijving:Single tool holster with retractor
- Environmentally Preferable:
- VPE:1
Specifications
About this item
Single tool harness holster with retractor.
- Adjusts to virtually any wrist size with hook and loop system
- Replaceable Steel cable retractor fits into a sewn-in pocket and allows tools to easily be tied off using an attached thumb-operated trigger
- Elastic band helps ensure a comfortable fit
- Attached trigger can be used to quickly work with many hand tools
The DBI SALA® Single tool holster for a harness comfortably accommodates hand tools in a compact, user-friendly design that is loaded with innovative product features like Hi Vis reflectors and a water drainage vent. Constructed from a premium heavy-duty ballistic denier fabric, these holsters are engineered to withstand punishing environments. Heavy-duty 25 mm (1") D-rings are built into both sides of the holster and are load rated for 2,3 kg (5 lbs.). A neoprene pocket on the front of the holster includes a retractor tether.
Designed for use in general industrial, oil and gas, mining, construction, and transportation industries.