Orders LinkContactUsLinkComponent
Fall protection harnesses, 1 point
Fall protection harnesses, 1 point
Harness, 0,88 kg
Catalogus # PRTWFP11RER
Leverancier:   Portwest
Fall protection harnesses, 1 point
Catalogus # PRTWFP11RER
Leverancier:  Portwest
Fall protection harnesses, 1 point
Fall protection harnesses, 1 point
Harness, 0,88 kg
Price
34,90 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
Fall Protection is put in place to prevent the risks associated with falling from heights, reducing impact force, restricting obstacle/ground collision and restricting users from fall hazard areas.
  • Dorsal D-ring
  • Chest and leg adjustment points
  • Flat bar buckles
  • Inspection cover for protection of product label
  • Retail box which aids presentation for retail sales
  • Made from polyester webbing, alloy steel, heat treated
Ideal for fall arrest or work restraint systems, our entry level full body harness is light weight and easy to use, allowing a secure fit.
Certifications: EN 361
Specificaties
  • Omschrijving:
    Harness
  • Gewicht:
    0,88 kg