Orders LinkContactUsLinkComponent
Fall protection harnesses, 2 point
Fall protection harnesses, 2 point
Harness, 1,2 kg, 2-punts
Catalog # PRTWFP13RER
Supplier:   Portwest
Fall protection harnesses, 2 point
Catalog # PRTWFP13RER
Supplier:  Portwest
Fall protection harnesses, 2 point
Fall protection harnesses, 2 point
Harness, 1,2 kg, 2-punts
Price
56,60 €
Log in for availability
1 * 1 ST
Product Details & Documents
Fall Protection is put in place to prevent the risks associated with falling from heights, reducing impact force, restricting obstacle/ground collision and restricting users from fall hazard areas.
  • Superior strength
  • Fully adjustable shoulder, chest and leg straps
  • Chest loops
  • Light weight and easy to use
  • Made from polyester webbing, alloy steel, heat treated
Features front and back D-rings and is widely used in the construction industry.
Certifications: EN 361
Specifications
  • Bevestigingspunten:
    2-punts
  • Omschrijving:
    Harness
  • Gewicht:
    1,2 kg