Multi-element solution 3 with Hg for ICP
Multi-Element Solution 3 with Hg, 5% HNO₃, Plastic bottle, 100 mL
Catalogus # HONCE2178-100ML
Leverancier: Honeywell Chemicals
Multi-element solution 3 with Hg for ICP
Catalogus # HONCE2178-100ML
Leverancier: Honeywell Chemicals
CAS-nummer:
Ga naar:
Product Details & Documents
Certified Reference Material (CRM).
10 mg/L each of Ag, Al, As, Ba, Be, Bi, Ca, Cd, Co, Cr, Cs, Cu, Fe, Ga, Hg, In, K, Li, Mg, Mn, Na, Ni, Pb, Rb, Se, Sr, Tl, U, V, Zn in HNO₃ 5%
Specificaties
Specificaties
- Matrix:5% HNO₃
- Verpakking:Plastic bottle
- VPE:100 mL
- Omschrijving:Multi-Element Solution 3 with Hg