Pocket refractormeter, IR Brix meter (Strawberry), PAL-Hikari 4
Catalogus # ATAG5454
Leverancier: Atago
Specificaties
- Omschrijving:Pocket IR Brix Meter PAL-Hikari 4 (Strawberry)
- Environmentally Preferable:
- VPE:1
Specificaties
Over dit artikel
Non-destructive strawberry Brix meter.
- The Brix (sugar level) can be measured by placing the fruit on the sample stage with no need of cutting or squeezing
It requires no more cumbersome wiping and cleansing after each measurement.
Bestelinformatie: Requires 3 ˣ AAA batteries.