Avantor® Hichrom HI-65 HT, GC Columns
Leverancier: Avantor
Certificaten
Over dit artikel
High Temperature column ideal for triglycerides separations based on carbon number and degree of unsaturation.
Max temperature limits may change depending on the film thickness.
Specificaties
- Catalog No:
- HI47-32-010-15
- HI47-32-010-30
- HI47-25-010-15
- HI47-25-010-30
- HIGCHI47-25-010-25
- HI47-53-010-30
- Phase name:
- HI-65 HT
- HI-65 HT
- HI-65 HT
- HI-65 HT
- HI-65 HT
- Phase composition:
- 65% Phenyl, 35% Methyl Polysiloxane - high temperature
- 65% Phenyl, 35% Methyl Polysiloxane - high temperature
- 65% Phenyl, 35% Methyl Polysiloxane - high temperature
- 65% Phenyl, 35% Methyl Polysiloxane - high temperature
- 65% Phenyl, 35% Methyl Polysiloxane - high temperature
- Polarity:
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Polar stationary phase
- Crossbond:
- Yes
- Yes
- Yes
- Yes
- Yes
- Temperatuurlimiet:
- 370 °C
- 370 °C
- 370 °C
- 370 °C
- 370 °C
Specificaties
Frequently Bought Together