Spill berm, QuickBerm® rigid lock, EG-5
Catalogus # BRDY198532
Leverancier: Brady
Specificaties
- Capaciteit:890 L
- Omschrijving:Spill berm, QuickBerm® rigid lock, EG-5
- b×d×h:1422×2642×305 mm
- Environmentally Preferable:
- VPE:1
Specificaties
Over dit artikel
The extreme durable Rigid-Lock QuickBerm® provided immediate, secure deployment and spill-readiness. The built-in Rigid-Lock wall supports maximise usable space inside the berm and greatly reduce any tripping hazard on the berm perimeter.
- Made of PVC coated fabric
- Black in colour
The high performance material withstands a broad range of chemicals including petroleum, oils, grease, and most acids and a broad UV resistance - durable enough for extended wear.