Face shield, DFS1
Catalog # 111-2494
Supplier: Bollé Safety
Specifications
- Omschrijving:DFS1 face shield
- l×b:220x330 mm
- VPE:1 KIT
Specifications
About this item
The DFS1 face shield perfectly protects against splashes and droplets. Designed to fit first reponders’ needs, DFS1 is simple to use, economic, protective, and can be worn over glasses.
- Protective band made from foam
- Anti-fog and blue protective shield coating
- Adapted to prescription eyewear
- Protects against blood and bodily fluids
- Elastic headband
Easy to assemble, the face shield is supplied as three separate components - an 0,2 mm thick PET visor with PU frame, foam band and white polyester strap.
The protective shield should be cleaned regularly. Pitted, scratched, or damaged shields should be replaced. The face shield must be stored in a clean and dry environment.