Order Entry
Belgium
ContactUsLinkComponent
 
N-Methyl-N-octylamine ≥98.0% (by GC, titration analysis)
  TCIAM2219-5G
 :  TCI
 :  
N-Methyl-N-octylamine ≥98.0% (by GC, titration analysis)
  TCIAM2219-5G
 :  TCI
 :  

 

  • VPE:
    5 g
  • Seq:
    3173

 

 

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %

 

 

 :
 :143.27 g/mol
 :51°C
 :63°C
 :219-462-1
 :2735
 :8,II
 :GHS05
 :2439-54-5
 :C₉H₂₁N
 :MFCD00048927