Deflazacort ≥98.0% (by HPLC, total nitrogen)
Catalogus # TCIAD4523-1G
Leverancier: TCI
CAS-nummer:
Specificaties
- VPE:1 g
- Seq:3047
Specificaties
Specificatie testresultaten
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (with Total Nitrogen):min. 98.0 %
- Melting point:256.0 to 260.0 deg-C
- Specific rotation [a]20/D:+63.0 to +74.0 deg (C=0.5, CHCl3)
Specificatie testresultaten
Over dit artikel
:
:441.52 g/mol
Merck index:14,02862
Temperatuur bij opslag:Koelkast
EINECS:238-483-7
CAS-nummer:14484-47-0
Formule:C₂₅H₃₁NO₆
MDL-nummer:MFCD00866106