Order Entry
Belgium
ContactUsLinkComponent
Cinnamyl chloride ≥95.0% (by GC)
Corrosive
Toxic
Cinnamyl chloride ≥95.0% (by GC)
Catalog # TCIAC1235-25G
Supplier:  TCI
CAS Number:  
Corrosive
Toxic
Cinnamyl chloride ≥95.0% (by GC)
Catalog # TCIAC1235-25G
Supplier:  TCI
CAS Number:  

Specifications

  • VPE:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 95.0 %

Specification Test Results

About this item

 :
 :152.62 g/mol
Boiling Pt:136°C
Flash Pt:79°C
EINECS:220-246-4
UN:2927
ADR:6.1,II
 :GHS05
 :GHS06
CAS Number:2687-12-9
Formula:C₉H₉Cl
MDL Number:MFCD00000986