
1-(3-Cyanopropyl)pyrrolidine ≥95.0% (by GC)
Catalog # TCIAC1231-25ML
Supplier: TCI
CAS Number:
Specifications
- VPE:25 mL
- Seq:2315
Specifications
Specification Test Results
- Appearance:Colorless to Light orange to Yellow clear liquid
- Purity (GC):min. 95.0 %
Specification Test Results
About this item
:
:138.21 g/mol
Boiling Pt:115°C
Flash Pt:99°C
EINECS:252-612-4
:GHS07
CAS Number:35543-25-0
Formula:C₈H₁₄N₂
MDL Number:MFCD00003183