Allolactose ≥98.0% (by HPLC)
Catalog # TCIAA2630-100MG
Supplier: TCI
CAS Number:
Specifications
- VPE:100 mg
- Seq:2440
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White powder to crystaline
- Purity (HPLC) (CAD):min. 98.0 area%
Specification Test Results
About this item
Synonyms: 6-O-beta-D-Galactopyranosyl-D-glucopyranose
:
:342.30 g/mol
Storage Temperature:Vriezer
CAS Number:28447-39-4
Formula:C₁₂H₂₂O₁₁
MDL Number:MFCD15145331
Melting Pt:168°C