VWR®, Titanium standard solution, 1000 mg/L Ti in water with hydrofluoric acid (max. 1%) (from (NH4)2TiF6) ARISTAR® standard for ICP
456262B
:
- Concentratie:1000 mg/L
- Matrix:H₂O/tr HF
- Verpakking:Plastic bottle
- VPE:100 mL
- Element:Titaan
- Symbool:Ti
(NH₄)₂ TiF₆ in H₂O tr. HF.
Traceable to SRM from NIST, tested in an ISO Guide 34 / ISO17025 accredited laboratory.
Supplied with certificate of analysis.