VWR® Gurr™ Histological Fixatives, AFA
81009.362
:
- Verpakking:Plastic container
- VPE:5 L
- Omschrijving:Histological fixative, AFA
- Seq:03
Acidified alcoholic formalin for the fixation of glycogen.
Contains acetic acid, ethanol and formaldehyde.