Quantitative filter papers, pyramid folded, Whatman™
Supplier: Whatman products (Cytiva)
Certificates
About this item
Easily fits into conical funnels.
- High quality and consistency
- Easy to use
Specifications
- Catalog No:
- WHAT989510116
- WHAT989610112
- WHAT989710112
- WHAT9891-128
- WHAT989810112
- WHAT9892-128
- WHAT1540-10123
- WHAT989810116
- WHAT1540-10124
- WHAT989510112
- Nr:
- 41
- 42
- 1
- 6
- 4
- 40
- 540
- 4
- 540
- 41
- Gewicht:
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- 85 g/m²
- Dikte:
- 215 µm
- 215 µm
- 215 µm
- 215 µm
- 215 µm
- 215 µm
- 215 µm
- 215 µm
- 215 µm
- 215 µm
- Asgehalte:
- 0,007%
- 0,007%
- 0,007%
- 0,007%
- 0,007%
- 0,007%
- 0,007%
- 0,007%
- 0,007%
- 0,007%
Specifications
Frequently Bought Together