Orders LinkContactUsLinkComponent
Affinity chromatography columns, HiTrap™ Capto™ AVB
 :   Cytiva
Affinity chromatography columns, HiTrap™ Capto™ AVB
 :  Cytiva
Affinity chromatography columns, HiTrap™ Capto™ AVB
HiTrap™ columns packed with Capto™ AVB affinity chromatography resin designed for the purification of adeno-associated virus (AAV). Capto™ AVB is based on a rigid base matrix, delivering excellent pressure-flow properties to AAV production.
  • Efficient purification of AAV of several subclasses by affinity chromatography
  • Prepacked HiTrap columns for easy screening and convenient process development, as well as small scale purification
  • High selectivity and excellent scalability
The possibility to run at higher flow rates and bed heights increases flexibility in process design and allows for an increased productivity.
  • Catalog No:
  • 17-3722-11
  • 17-3722-12
  • Max. druk:
  • 5 bar
  • 5 bar
  • Opslag:
  • 4 to 8 °C, 20% ethanol
  • 4 to 8 °C, 20% ethanol
Product Family Options
  • Omschrijving
    Kolombehuizing
    Bedafmetingen
    Capaciteit
    Stroomsnelheid
  • HiTrap™ Capto™ AVB
    PP
    7×25 mm
    1 ml
    <4 ml/min
    1 * 5 ST
    1 * 5 ST - 1 980,00 €
    1 980,00 €
    1 * 5 ST
       1 980,00 €
    Specifications
    Cat. No.
    17-3722-11
    Max. druk
    5 bar
    Opslag
    4 to 8 °C, 20% ethanol
  • HiTrap™ Capto™ AVB
    PP
    16×25 mm
    5 ml
    <20 ml/min
    1 * 1 ST
    1 * 1 ST - 1 930,00 €
    1 930,00 €
    1 * 1 ST
       1 930,00 €
    Specifications
    Cat. No.
    17-3722-12
    Max. druk
    5 bar
    Opslag
    4 to 8 °C, 20% ethanol