General purpose gloves, Showa 370
Supplier: Showa
Certificates
About this item
These lightweight palm-coated nylon gloves are ideal for protecting hands against fluids such as oils. They allow finger dexterity to be maintained whilst being robust and hard-wearing.
- Nitrile coating protects the hand from oil, hydrocarbon and grease penetration
- Breathable back of hand and fingers
- High levels of dexterity and tactility
- Seamless 13 gauge lint-free liner provides excellent dry grip and resistant to deformation
Applications include assembling components, handling greasy components, inspection, machining and maintenance.These gloves are ideally suited for aerospace, agriculture, automotive, construction, logistics and manufacturing.
Certifications: EN 388:2016 (4121X); Cat. II
Packaging: 10 pairs per pack.
Caution: Do not use in cases of chemical, electrical or thermal risk, Store away from light and humidity and wash at 40 °C max, using a neutral detergent.
Specifications
- Catalog No:
- 112-5801
- 112-5800
- 112-5799
- 112-5798
- 112-5797
- 112-5802
- Model:
- Showa 370
- Showa 370
- Showa 370
- Showa 370
- Showa 370
- Showa 370
- Ambidexter/handspecifiek:
- Handspecifiek
- Handspecifiek
- Handspecifiek
- Handspecifiek
- Handspecifiek
- Handspecifiek
- Voor eenmalig gebruik/herbruikbaar:
- Herbruikbaar
- Herbruikbaar
- Herbruikbaar
- Herbruikbaar
- Herbruikbaar
- Herbruikbaar
- Voeringmateriaal:
- Nylon
- Nylon
- Nylon
- Nylon
- Nylon
- Nylon
- Materiaal:
- Nitril
- Nitril
- Nitril
- Nitril
- Nitril
- Nitril
- Afwerking:
- Palm coated
- Palm coated
- Palm coated
- Palm coated
- Palm coated
- Palm coated
- Steken per inch (Gauge):
- 13
- 13
- 13
- 13
- 13
- 13
- Manchetstijl:
- Gebreide manchet
- Gebreide manchet
- Gebreide manchet
- Gebreide manchet
- Gebreide manchet
- Gebreide manchet
- EN-norm:
- EN 388:2016 (4121X)
- EN 388:2016 (4121X)
- EN 388:2016 (4121X)
- EN 388:2016 (4121X)
- EN 388:2016 (4121X)
- EN 388:2016 (4121X)
Specifications
Frequently Bought Together