ISA (ionic strength adjustment solution: 1 M KCl), Supelco®
ISA (ionic strength adjustment solution), 1 M KCl, Plastic bottle, 500 mL
Supplier: Merck
ISA (ionic strength adjustment solution: 1 M KCl), Supelco®
Catalog # 58221-500ML-F
Supplier: Merck
CAS Number:
Jump to:
Product Details & Documents
Solution used to establish a constant ionic strength in a sample solution containing e.g. calcium for det. with ISE.
- Prepared with high purity salts
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Verpakking:Plastic bottle
- VPE:500 mL
- Omschrijving:ISA (ionic strength adjustment solution)
- Bestanddelen:1 M KCl
- Cat. No.:58221-500ML-F
- M-Clarity quality level:MQ100
Specifications