Control units
Supplier: 2MAG
About this item
4 Options Available Below
Product Details & Documents
Specifications for Product Family
Specifications
- Catalog No:
- 442-0769
- 442-0768
- 442-0770
- TMAG90210_CH
- TMAG90210_EU
- TMAG90210_UK
- 442-0772
- 442-0771
- 442-0773
- TMAG90410_CH
- TMAG90410_EU
- TMAG90410_UK
- Model:
- MIXcontrol 20
- MIXcontrol 20
- MIXcontrol 20
- MIXcontrol 20
- MIXcontrol 20
- MIXcontrol 20
- MIXcontrol 40
- MIXcontrol 40
- MIXcontrol 40
- MIXcontrol 40
- MIXcontrol 40
- MIXcontrol 40
- Toerentalbereik:
- 100 - 1600 min⁻¹
- 100 - 1600 min⁻¹
- 100 - 1600 min⁻¹
- 100 - 1600 min⁻¹
- 100 - 1600 min⁻¹
- 100 - 1600 min⁻¹
- 100 - 2000 min⁻¹
- 100 - 2000 min⁻¹
- 100 - 2000 min⁻¹
- 100 - 2000 min⁻¹
- 100 - 2000 min⁻¹
- 100 - 2000 min⁻¹
- Voeding:
- 20 W
- 20 W
- 20 W
- 20 W
- 20 W
- 20 W
- 40 W
- 40 W
- 40 W
- 40 W
- 40 W
- 40 W
- Gewicht:
- 1,9 kg
- 1,9 kg
- 1,9 kg
- 1,9 kg
- 1,9 kg
- 1,9 kg
- 1,9 kg
- 1,9 kg
- 1,9 kg
- 1,9 kg
- 1,9 kg
- 1,9 kg
- b×d×h:
- 200×155×38 mm
- 200×155×38 mm
- 200×155×38 mm
- 200×155×38 mm
- 200×155×38 mm
- 200×155×38 mm
- 200×155×48 mm
- 200×155×48 mm
- 200×155×48 mm
- 200×155×48 mm
- 200×155×48 mm
- 200×155×48 mm
Specifications
Product Family Options
Product Information
- OmschrijvingAvailabilityPrice
- MIXcontrol 20, CH-stekkerDiscontinued-Specifications:
More
Specifications
Cat. No.442-0769ModelMIXcontrol 20Toerentalbereik100 - 1600 min⁻¹Voeding20 WGewicht1,9 kgb×d×h200×155×38 mm - Specifications:
More
Specifications
Cat. No.442-0768ModelMIXcontrol 20Toerentalbereik100 - 1600 min⁻¹Voeding20 WGewicht1,9 kgb×d×h200×155×38 mm - MIXcontrol 20, UK-stekkerDiscontinued-Specifications:
More
Specifications
Cat. No.442-0770ModelMIXcontrol 20Toerentalbereik100 - 1600 min⁻¹Voeding20 WGewicht1,9 kgb×d×h200×155×38 mm - MIXcontrol 20, met RS232 interface, CH-stekkerDiscontinued-Specifications:
More
Specifications
Cat. No.TMAG90210_CHModelMIXcontrol 20Toerentalbereik100 - 1600 min⁻¹Voeding20 WGewicht1,9 kgb×d×h200×155×38 mm - Specifications:
More
Specifications
Cat. No.TMAG90210_EUModelMIXcontrol 20Toerentalbereik100 - 1600 min⁻¹Voeding20 WGewicht1,9 kgb×d×h200×155×38 mm - MIXcontrol 20, met RS232 interface, UK-stekkerDiscontinued-Specifications:
More
Specifications
Cat. No.TMAG90210_UKModelMIXcontrol 20Toerentalbereik100 - 1600 min⁻¹Voeding20 WGewicht1,9 kgb×d×h200×155×38 mm - MIXcontrol 40, CH-stekkerDiscontinued-Specifications:
More
Specifications
Cat. No.442-0772ModelMIXcontrol 40Toerentalbereik100 - 2000 min⁻¹Voeding40 WGewicht1,9 kgb×d×h200×155×48 mm - Specifications:
More
Specifications
Cat. No.442-0771ModelMIXcontrol 40Toerentalbereik100 - 2000 min⁻¹Voeding40 WGewicht1,9 kgb×d×h200×155×48 mm - MIXcontrol 40, UK-stekkerDiscontinued-Specifications:
More
Specifications
Cat. No.442-0773ModelMIXcontrol 40Toerentalbereik100 - 2000 min⁻¹Voeding40 WGewicht1,9 kgb×d×h200×155×48 mm - MIXcontrol 40, met RS232 interface, CH-stekkerDiscontinued-Specifications:
More
Specifications
Cat. No.TMAG90410_CHModelMIXcontrol 40Toerentalbereik100 - 2000 min⁻¹Voeding40 WGewicht1,9 kgb×d×h200×155×48 mm - Specifications:
More
Specifications
Cat. No.TMAG90410_EUModelMIXcontrol 40Toerentalbereik100 - 2000 min⁻¹Voeding40 WGewicht1,9 kgb×d×h200×155×48 mm - MIXcontrol 40, met RS232 interface, UK-stekkerDiscontinued-Specifications:
More
Specifications
Cat. No.TMAG90410_UKModelMIXcontrol 40Toerentalbereik100 - 2000 min⁻¹Voeding40 WGewicht1,9 kgb×d×h200×155×48 mm