Avantor® Hichrom, Tubing Cutter for Polymer Tubing
Catalog # HICHHI-191
Supplier: Avantor
Specifications
- Omschrijving:Standard polymer tubing cutter
- VPE:1
Specifications
About this item
Standard cutter for 1/16" and 1/8" O.D. tubing.
- Compatible with PEEK and PTFE tubing
- Flat burr-free cuts
- Guide holes to ensure precise cutting
Delivery: Includes five replacement blades.